Skip to Content
Merck
All Photos(1)

Key Documents

33154

Sigma-Aldrich

Dithizone

puriss. p.a., reag. Ph. Eur., ≥98% (chelatometric), for metal titration

Synonym(s):

Diphenylthiocarbazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNHCSN=NC6H5
CAS Number:
Molecular Weight:
256.33
Beilstein:
748838
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
reag. Ph. Eur.

Quality Level

grade

puriss. p.a.

Assay

≥98% (chelatometric)

technique(s)

titration: suitable

impurities

≤0.001% heavy metals (as Pb)

ign. residue

≤0.2% (as SO4)

mp

168 °C (dec.) (lit.)

absorption

≥1450 at 605 nm

SMILES string

S=C(NNc1ccccc1)\N=N\c2ccccc2

InChI

1S/C13H12N4S/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14H,(H,16,18)/b17-15+

InChI key

UOFGSWVZMUXXIY-BMRADRMJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dithizone (Diphenylthiocarbazone) is used as chelating agent for zinc. It is widely used for the liquid-liquid and solid phase extraction of various metals since it forms stable complexes with the metals.

Application

Dithizone (Diphenylthiocarbazone) may be used for the in vitro characterization of differentiated pancreatic islets clusters in differentiated embryonic stem cell cultures, as it forms chelates with zinc present in huge quantities in pancreatic islets, thereby selectively staining them. It may be used to form complex with silver ions in the cloud point preconcentration step before the trace level quantification of silver by flame atomic absorption spectrometry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Shiroi et al.
Stem cells (Dayton, Ohio), 20(4), 284-292 (2002-07-12)
Embryonic stem (ES) cells have a pluripotent ability to differentiate into a variety of cell lineages in vitro. We have recently identified the emergence of cellular clusters within differentiated ES cell cultures by staining with dithizone (DTZ). DTZ is a
Selective cloud point extraction and preconcentration of trace amounts of silver as a dithizone complex prior to flame atomic absorption spectrometric determination.
Manzoori JL and Karim-Nezhad G.
Analytica Chimica Acta, 484(2), 155-161 (2003)
Yong-guang Yin et al.
Talanta, 81(4-5), 1788-1792 (2010-05-06)
A novel and simple solid phase extraction (SPE)-high performance liquid chromatography (HPLC)-inductively coupled plasma mass spectrometry (ICP-MS) method was developed for determination of inorganic mercury (IHg), methylmercury MeHg and ethylmercury (EtHg) in water samples in the present work. The procedure
E Karaoz et al.
Cell death & disease, 1, e36-e36 (2011-03-03)
The maintenance of viable and functional islets is critical in successful pancreatic islet transplantation from cadaveric sources. During the isolation procedure, islets are exposed to a number of insults including ischemia, oxidative stress and cytokine injury that cause a reduction
Xiaodong Wen et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 782-787 (2012-08-21)
In this work, a new coupling of spectrophotometric determination with preconcentration method named as ultrasound-assisted emulsification dispersive liquid-liquid microextraction (UAE-DLLME) for trace silver was firstly established. Disperser solvent in traditional DLLME was substituted by ultrasound-assisted emulsification, which could afford more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service