Skip to Content
Merck
All Photos(1)

Documents

32977

Supelco

Terbutryn-d5

PESTANAL®, analytical standard

Synonym(s):

N-tert-Butyl-N′-ethyl-d5-6-methylthio-1,3,5-triazine-2,4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D5H14N5S
Molecular Weight:
246.39
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

[2H]C([2H])(C([2H])([2H])[2H])NC1=NC(SC)=NC(NC(C)(C)C)=N1

InChI

1S/C10H19N5S/c1-6-11-7-12-8(15-10(2,3)4)14-9(13-7)16-5/h6H2,1-5H3,(H2,11,12,13,14,15)/i1D3,6D2

InChI key

IROINLKCQGIITA-YRYIGFSMSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service