Skip to Content
Merck
All Photos(1)

Key Documents

32759

Supelco

Patulin

reference material

Synonym(s):

4-Hydroxy-4H-furo[3,2-c]pyran-2(6H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O4
CAS Number:
Molecular Weight:
154.12
Beilstein:
149675
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
reference material

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

OC1OCC=C2OC(=O)C=C12

InChI

1S/C7H6O4/c8-6-3-4-5(11-6)1-2-10-7(4)9/h1,3,7,9H,2H2

InChI key

ZRWPUFFVAOMMNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Patulin (PAT) is a mycotoxin of the polyketide lactone type, usually produced by species of the Penicillium, Aspergillus and Byssochlamys genera.

Application

PAT may be used as a reference standard for the determination of patulin in:
  • Fruit-based products by QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) method and solid phase extraction (SPE) followed by analysis using ultra-high performance liquid chromatography combined with electrospray ionization tandem mass spectrometry (UHPLC-ESI-MS/MS).
  • Apple juice samples by single drop liquid-liquid-liquid microextraction (SD-LLLME) followed by LC-MS/MS analysis.
  • Model food matrices by “dilute and shoot” LC-MS/MS.
  • Dairy products by QuEChERS procedure and UHPLC-ESI quadrupole orbitrap (Q-Orbitrap) mass spectrometry.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecularly imprinted polymer for extraction of patulin in apple juice samples.
Anene A, et al.
Food Control, 70, 90-95 (2016)
Multi-mycotoxin analysis in dairy products by liquid chromatography coupled to quadrupole orbitrap mass spectrometry.
Jia W, et al.
Journal of Chromatography A, 1345, 107-114 (2014)
Optimization and validation of a quantitative liquid chromatography-tandem mass spectrometric method covering 295 bacterial and fungal metabolites including all regulated mycotoxins in four model food matrices.
Malachova A, et al.
Journal of Chromatography A, 1362, 145-156 (2014)
Monitoring survey of patulin in a variety of fruit-based products using a sensitive UHPLC-MS/MS analytical procedure.
Vaclavikova M, et al.
Food Control, 47, 577-584 (2015)
Determination of patulin in apple juice by single-drop liquid-liquid-liquid microextraction coupled with liquid chromatography-mass spectrometry.
Li X, et al.
Food Chemistry, 257, 1-6 (2018)

Related Content

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service