Skip to Content
Merck
All Photos(2)

Documents

32528

Supelco

Pethoxamid

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-N-(2-ethoxyethyl)-N-(2-methyl-1-phenyl-1-propenyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22ClNO2
CAS Number:
Molecular Weight:
295.80
Beilstein:
4873772
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOCCN(C(=O)CCl)C(=C(/C)C)\c1ccccc1

InChI

1S/C16H22ClNO2/c1-4-20-11-10-18(15(19)12-17)16(13(2)3)14-8-6-5-7-9-14/h5-9H,4,10-12H2,1-3H3

InChI key

CSWIKHNSBZVWNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pethoxamid is a herbicide belonging to the class of chloro-acetamides. It is primarily used for weed control management in crop plantations.

Application

Pethoxamid may be used as a reference standard in the determination of pethoxamid in water samples and commercial formulations using electrochemical technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

First electrochemistry of herbicide pethoxamid and its quantification using electroanalytical approach from mixed commercial product.
Jevtic S, et al.
Electrochimica Acta, 277, 136-142c-136-142c (2018)
Effect of precipitation on the dissipation, efficacy and selectivity of three chloroacetamide herbicides in sunflower.
Jursik M, et al.
Plant, Soil and Environment, 59(4), 175-182 (2013)
Kaan Georg Kutlucinar et al.
Electrophoresis, 42(4), 490-500 (2020-12-18)
Nontargeted analysis of water samples using liquid chromatography combined with high-resolution mass spectrometers is an emerging approach for surface water monitoring and evaluation of water treatment processes. In this study, sample preconcentration via direct, large volume injection with 500 μL and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service