Skip to Content
Merck
All Photos(1)

Key Documents

32396

Supelco

Amicarbazone

PESTANAL®, analytical standard

Synonym(s):

4-Amino-N-tert-butyl-4,5-dihydro-3-isopropyl-5-oxo-1,2,4-1H-triazole-1-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$762.00

R$762.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
R$762.00

About This Item

Empirical Formula (Hill Notation):
C10H19N5O2
CAS Number:
Molecular Weight:
241.29
Beilstein:
12242812
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$762.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

NN1C(C(C)C)=NN(C(NC(C)(C)C)=O)C1=O

InChI

1S/C10H19N5O2/c1-6(2)7-13-15(9(17)14(7)11)8(16)12-10(3,4)5/h6H,11H2,1-5H3,(H,12,16)

InChI key

ORFPWVRKFLOQHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amicarbazone is a photosystem II (PSII)-inhibiting herbicide that belongs to the triazolinone herbicide family,[1] with a broad spectrum of weed control.[2]

Application

Amicarbazone may be used as a reference standard for the determination of amicarbazone and its metabolite residues in plants by a modified QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) method by liquid chromatography-tandem mass spectrometry.[3]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annual bluegrass (Poa annua) control in creeping bentgrass putting greens with amicarbazone and paclobutrazol.
Jeffries M D, et al.
Weed Technology, 27(3), 520-526 (2013)
Determination of Amicarbazone and Its Metabolites in Corn and Its Plant with a Modified QuEChERS by Liquid Chromatography-Tandem Mass Spectrometry.
Mao-Feng D, et al.
Chinese Journal of Analytical Chemistry, 43(5), 663-668 (2015)
Photochemical fate of amicarbazone in aqueous media: laboratory measurement and simulations.
Silva M P, et al.
Environmental Engineering Science, 32(8), 730-740 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service