Skip to Content
Merck
All Photos(1)

Key Documents

311456

Sigma-Aldrich

Sodium peroxide

ACS reagent, ≥93.0%

Synonym(s):

Sodium superoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O2
CAS Number:
Molecular Weight:
77.98
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥93.0%
grade:
ACS reagent
form:
beads
powder
Pricing and availability is not currently available.

grade

ACS reagent

Assay

≥93.0%

form

beads
powder

reaction suitability

reagent type: oxidant

impurities

≤0.003% N compounds

pH

12.8 (20 °C, 100 g/L)

mp

460 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.001%

cation traces

Fe: ≤0.005%
heavy metals (as Pb): ≤0.002%

SMILES string

[Na+].[Na+].[O-][O-]

InChI

1S/2Na.O2/c;;1-2/q2*+1;-2

InChI key

PFUVRDFDKPNGAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Calvert et al.
The American journal of clinical nutrition, 49(5), 901-903 (1989-05-01)
A safer method of oxidation of Cr2O3-containing fecal samples from transit-time studies was developed using sodium peroxide to replace perchloric acid as the oxidizing agent. The percentage recovery of Cr2O3 with this method was compared with that of perchloric acid
[Rapid ultraviolet-absorption photometric determination of the protein content of aqueous extracts in the microgram area using Na2O2].
B Löhrke
Zeitschrift fur medizinische Laboratoriumsdiagnostik, 27(2), 92-95 (1986-01-01)
Christine Sedgley
Journal of the history of dentistry, 52(2), 61-65 (2004-08-06)
The concept of the germ theory of disease combined with the development of dentistry during the latter half of the 19th century had a direct effect on the practice of endodontics. The significance of root canal irrigation to endodontics strengthened
Tianji Li et al.
Environmental science & technology, 39(24), 9665-9668 (2006-02-16)
We tested a mixture of NaOH and carbon and found it to react rapidly with NO gas, thus demonstrating a promising new method of flue gas denitrification (FGD). We determined the reaction products: gas phases were analyzed by GC-MS and
K J Sreeram et al.
Journal of environmental monitoring : JEM, 3(5), 526-530 (2001-11-07)
The processing of chromite ore is associated with the generation of large quantities of solid wastes containing chromium, which have been disposed of as landfill for many years. The mobilization and operational speciation of chromium contained in soils contaminated with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service