Skip to Content
Merck
All Photos(1)

Key Documents

31115

Supelco

Methoprotryne

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21N5OS
CAS Number:
Molecular Weight:
271.38
Beilstein:
616626
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COCCCNc1nc(NC(C)C)nc(SC)n1

InChI

1S/C11H21N5OS/c1-8(2)13-10-14-9(12-6-5-7-17-3)15-11(16-10)18-4/h8H,5-7H2,1-4H3,(H2,12,13,14,15,16)

InChI key

DDUIUBPJPOKOMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methoprotryne is a triazine herbicide intended to control annual and perennial grass in a variety of crops. Triazines herbicides act by inhibiting photosynthesis in all organisms with oxygen-evolving photosystems.

Application

Methoprotryne may be used as a reference standard for the determination of the analyte:
  • In processed cereal samples from Chinese total diet study (TDS) using selective and sensitive isotope dilution–high performance liquid chromatography–linear ion trap mass spectrometry (Isotope Dilution–HPLC–LIT-MS3).
  • In drinking water by a novel dispersive micro solid phase extraction (DMSPE) method based on a polymer cation exchange material (PCX) with ultrahigh-performance liquid chromatography?high-resolution mass spectrometric detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of 19 triazine pesticides and degradation products in processed cereal samples from Chinese total diet study by isotope dilution-high performance liquid chromatography-linear ion trap mass spectrometry.
Li P, et al.
Analytica Chimica Acta, 781(1), 63-71 (2013)
The mode of action of triazine herbicides in plants.
Trebst A.
The Triazine Herbicides, 50(1), 101- 110 (2008)
Determination of triazine herbicides in drinking water by dispersive micro solid phase extraction with ultrahigh-performance liquid chromatography-high-resolution mass spectrometric detection.
Chen D, et al.
Journal of Agricultural and Food Chemistry, 63(44), 9855-9862 (2015)
Determination of triazine pesticides and related compounds in environmental water by liquid chromatography-mass spectrometry.
Tanabe A and Kawata K
Analytical Sciences, 20(1), 227- 230 (2004)
Xiaochun Wang et al.
Bulletin of environmental contamination and toxicology, 105(6), 899-905 (2020-11-21)
The aim of the current study was to track the composition, spatial and temporal distribution charactistics of triazine herbicides in arable soils and corns in typical agricultural regions of Liaoning Province, China. All samples were analyzed using high performance liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service