Skip to Content
Merck
All Photos(1)

Key Documents

30977

Supelco

6-Ring NOGE (Novolac glycidyl ether) mixture of isomers, chain-like or branched

analytical standard

Synonym(s):

6-Ring NOGE (Novolac glycidyl ether) mixture of isomers, chain-like or branched, NOGE 6-Ring (Novolac glycidyl ether) mixture of isomers, Novolac glycidyl ether 6-Ring

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
R$10,294.00

R$10,294.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
R$10,294.00

About This Item

Empirical Formula (Hill Notation):
C59H60O12
Molecular Weight:
961.10
UNSPSC Code:
85151701
NACRES:
NA.24

R$10,294.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥80.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service