293121
Thionyl chloride solution
2.0 M in methylene chloride
About This Item
Recommended Products
grade
for analytical purposes
Quality Level
vapor pressure
97 mmHg ( 20 °C)
form
liquid
concentration
2.0 M in methylene chloride
density
1.373 g/mL at 25 °C
SMILES string
ClS(Cl)=O
InChI
1S/Cl2OS/c1-4(2)3
InChI key
FYSNRJHAOHDILO-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Supplementary Hazards
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service