Skip to Content
Merck
All Photos(1)

Key Documents

29100

Sigma-Aldrich

Cyclohexanol

puriss., ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
906744
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.5 (vs air)

vapor pressure

0.98 mmHg ( 25 °C)

grade

puriss.

Assay

≥99.0% (GC)

autoignition temp.

572 °F

expl. lim.

12.25 %

impurities

≤0.5% cyclohexanone

refractive index

n20/D 1.465 (lit.)
n20/D 1.466

bp

160-161 °C (lit.)

mp

20-22 °C (lit.)

density

0.948 g/mL at 25 °C (lit.)

SMILES string

OC1CCCCC1

InChI

1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2

InChI key

HPXRVTGHNJAIIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup - DIN 51794

Flash Point(C)

64 °C - closed cup - DIN 51794

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 511-511 (1994)
Catalytic oxidation of cyclohexane to cyclohexanol and cyclohexanone over Co3O4 nanocrystals with molecular oxygen.
Zhou L, et al.
Applied Catalysis A: General, 292, 223-228 (2005)
Eagleson M.
Concise Encyclopedia Chemistry, 811-811 (1994)
Kinetic model of 2-cyclohexenone formation from cyclohexanol and 2-cyclohexenol dehydrogenation.
Simon E, et al.
Chemical Engineering Journal, 192, 129-137 (2012)
Controllable oxidation of cyclohexane to cyclohexanol and cyclohexanone by a nano-MnO x/Ti electrocatalytic membrane reactor.
Fang X, et al.
J. Catal., 329, 187-194 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service