Skip to Content
Merck
All Photos(1)

Key Documents

28100

Sigma-Aldrich

Crotyl bromide

technical, mixture of cis- and trans-isomers (~1:5), cis+trans, ~85% (GC)

Synonym(s):

trans-1-Bromo-2-butene

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
CH3CH=CHCH2Br
CAS Number:
Molecular Weight:
135.00
Beilstein:
1361394
MDL number:
UNSPSC Code:
12352100

grade

technical

Assay

~85% (GC)

contains

silver wool as stabilizer

impurities

~15% 3-bromo-1-butene

refractive index

n20/D 1.480 (lit.)
n20/D 1.480

bp

97-99 °C (lit.)

density

1.312 g/mL at 25 °C (lit.)

SMILES string

[H]\C(C)=C(\[H])CBr

InChI

1S/C4H7Br/c1-2-3-4-5/h2-3H,4H2,1H3/b3-2+

InChI key

AVMHMVJVHYGDOO-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service