Skip to Content
Merck
All Photos(1)

Key Documents

27827

Supelco

Poly(vinyl chloride-co-vinyl acetate-co-vinyl alcohol)

Selectophore, 91% vinyl chloride – 3% vinyl acetate – 6% vinyl alcohol

Synonym(s):

Copolymer of vinyl chloride, vinyl acetate and vinyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

powder

mol wt

Mr ~24500

General description

Visit our Sensor Applications portal to learn more.

Application

Poly(vinyl chloride-co-vinyl acetate-co-vinyl alcohol) was used in SPR (surface plasmon resonance) system, which was based on the wavelength shift, and was used for uranyl ion detection.[1]

Other Notes

Copolymer suitable for the preparation of membranes for ion-selective electrodes[2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uranyl ion detection based on wavelength-resolved surface plasmon resonance spectroscopy.
Chen H
Sensors and Actuators B, Chemical, 134 (2), 419-422 (2008)
Elimination of the asymmetry in neutral-carrier-based solvent polymeric membranes induced by proteins.
L F Dürselen et al.
Analytical chemistry, 60(14), 1455-1458 (1988-07-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service