Skip to Content
Merck
All Photos(1)

Documents

25345

Sigma-Aldrich

6-Chloronicotinic acid

purum, ≥97.0% (T)

Synonym(s):

6-Chloropyridine-3-carboxylic acid, 6-Chloronicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClNO2
CAS Number:
Molecular Weight:
157.55
Beilstein:
115993
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (T)

mp

190 °C (dec.) (lit.)

SMILES string

OC(=O)c1ccc(Cl)nc1

InChI

1S/C6H4ClNO2/c7-5-2-1-4(3-8-5)6(9)10/h1-3H,(H,9,10)

InChI key

UAWMVMPAYRWUFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehmet Karabacak et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(3), 876-883 (2008-03-25)
The experimental and theoretical study on the structures and vibrations of 6-chloronicotinic acid (6-CNA, C(6)H(4)ClNO(2)) are presented. The Fourier transform infrared spectra (4,000-50 cm(-1)) and the Fourier transform Raman spectra (3,500-50 cm(-1)) of the title molecule in solid phase have
A Segura Carretero et al.
Journal of chromatography. A, 1003(1-2), 189-195 (2003-08-06)
A method is described for the analysis of the insecticide imidacloprid [1-(6-chloro-3-pyridylmethyl)-N-nitroimidazolidin-2-ylideneamine] and its metabolite 6-chloronicotinic acid by micellar electrokinetic chromatography with diode-array detection at 270 and 227 nm, respectively. The best results were obtained using sodium dodecyl sulphate at
Gretty Ettiene et al.
Electrophoresis, 33(19-20), 2969-2977 (2012-09-22)
A sensitive and reliable method based on MEKC has been developed and validated for trace determination of neonicotinoid insecticides (thiamethoxam, acetamiprid, and imidacloprid) and the metabolite 6-chloronicotinic acid in water and soil matrices. Optimum separation of the neonicotinoid insecticides was
Won-Bo Shim et al.
Journal of agricultural and food chemistry, 57(3), 791-796 (2009-01-15)
A fluorescence polarization immunoassay (FPIA) for the quantitative determination of 6-chloronicotinic acid (6-CNA) using polyclonal antibody was developed. The 6-CNA-protein (bovine serum albumin and soybean trypsin inhibitor) conjugates and fluorescein-labeled 6-CNA derivative (tracer) were prepared and used as the immunogens
Kumiko Taira et al.
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology, 24(3), 222-230 (2011-09-29)
Neonicotinoid is a recently developed insecticide with worldwide use that has been increasing. It acts as a nicotinic acetylcholine receptor agonist. Chloropyridinyl neonicotinoid is a subgroup of neonicotinoid, and are commercially available as imidacloprid, nitenpyram, acetamiprid, and thiacloprid. The maximum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service