Skip to Content
Merck
All Photos(1)

Key Documents

239291

Sigma-Aldrich

Sodium dichromate dihydrate

ReagentPlus®, 99%

Synonym(s):

Sodium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2Cr2O7 · 2H2O
CAS Number:
Molecular Weight:
298.00
EC Number:
MDL number:
UNSPSC Code:
12352300

product line

ReagentPlus®

Assay

99%

reaction suitability

reagent type: oxidant

mp

91 °C (lit.)

SMILES string

O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2Na.2H2O.7O/h;;;;2*1H2;;;;;;;/q;;2*+1;;;;;;;;2*-1

InChI key

JYDRNIYTFCBIFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chad M Thompson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 123(1), 58-70 (2011-06-30)
Chronic ingestion of high concentrations of hexavalent chromium [Cr(VI)] in drinking water induces intestinal tumors in mice. To investigate the mode of action (MOA) underlying these tumors, a 90-day drinking water study was conducted using similar exposure conditions as in
Chad M Thompson et al.
Regulatory toxicology and pharmacology : RTP, 64(1), 68-76 (2012-06-19)
In vitro studies on hexavalent chromium [Cr(VI)] indicate that reduced forms of this metal can interact with DNA and cause mutations. Recently, Cr(VI) was shown to induce intestinal tumors in mice; however, Cr(VI) elicited redox changes, cytotoxicity and hyperplasia -
Isabelle Hininger et al.
Free radical biology & medicine, 42(12), 1759-1765 (2007-05-22)
Several studies have demonstrated beneficial effects of supplemental trivalent Cr in subjects with reduced insulin sensitivity with no documented signs of toxicity. However, recent studies have questioned the safety of supplemental trivalent Cr complexes. The objective of this study was
M O Elshazly et al.
PloS one, 10(6), e0126780-e0126780 (2015-06-02)
This study was conducted to investigate the possible protective effect of Spirulina platensis against chromium-induced nephrotoxicity. A total of 36 adult male Sprague-Dawley rats were divided into 4 equal groups (Gps). Gp1 served as control, rats of Gps 2, 3
Silvio De Flora et al.
Mutation research, 610(1-2), 38-47 (2006-07-29)
Chromium(VI) compounds are genotoxic in a variety of cellular systems. Their potential carcinogenicity is affected by toxicokinetic patterns restricting bioavailability to certain targets, and by metabolic pathways affecting interaction of chromate-derived reactive species with DNA. Epidemiological data indicate that chromium(VI)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service