Skip to Content
Merck
All Photos(1)

Key Documents

21300

Sigma-Aldrich

(+)-Camphor

purum, natural, ≥97.0% (sum of enantiomers, GC)

Synonym(s):

(1R)-(+)-Camphor, (1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2042745
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

grade

purum

Assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]20/D +42.5±2.5°, c = 10% in ethanol

quality

natural

mp

176-180 °C

SMILES string

[H][C@@]12CC[C@@](C)(C(=O)C1)C2(C)C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

(+)-Camphor, a chiral pool compound, has found extensive use as chiral building block; Many natural products have thus been prepared stereoselectively: a large number of compounds with a norbornane skeleton and other compounds

Application

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Target Organs

Lungs

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I. Kitagawa et al.
Chemical & Pharmaceutical Bulletin, 32, 1294-1294 (1984)
J.H. Hutchinson et al.
Canadian Journal of Chemistry, 65, 1-1 (1987)
Weiling Wang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(5), 6185-6196 (2019-02-16)
Human autosomal dominant polycystic kidney disease (ADPKD) is characterized by bilateral renal cysts that lead to a decline in kidney function. Previous studies reported aquaporin (AQP)-3 expression in cysts derived from collecting ducts in ADPKD. To study the role of
Tamera J Corte et al.
American journal of respiratory and critical care medicine, 190(2), 208-217 (2014-06-18)
Pulmonary hypertension (PH) associated with fibrotic idiopathic interstitial pneumonia (IIP; idiopathic pulmonary fibrosis and nonspecific interstitial pneumonia) confers important additional morbidity and mortality. To evaluate the safety and clinical efficacy of the dual endothelin-1 receptor antagonist bosentan in this patient
Camphor: a chiral starting material in natural product synthesis.
T Money
Natural product reports, 2(3), 253-289 (1985-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service