Skip to Content
Merck
All Photos(1)

Documents

19922

Sigma-Aldrich

tert-Butyldiphenylsilyl cyanide

≥97.0% (GC)

Synonym(s):

tert-Butyl-cyano-diphenylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NSi
CAS Number:
Molecular Weight:
265.42
Beilstein:
5907340
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

0.71 psi ( 55 °C)

Assay

≥97.0% (GC)

refractive index

n20/D 1.559 (lit.)
n20/D 1.560

bp

136 °C/0.7 mmHg (lit.)

density

1.017 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Si](C#N)(c1ccccc1)c2ccccc2

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for preparing silylated cyanohydrins from hindered ketones

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Golinski et al.
The Journal of Organic Chemistry, 58, 159-159 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service