Skip to Content
Merck
All Photos(1)

Key Documents

19115

Supelco

Silver ionophore VI

Selectophore, function tested

Synonym(s):

2-(Octadecyloxymethyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H43NO
CAS Number:
Molecular Weight:
361.60
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

quality

function tested

SMILES string

CCCCCCCCCCCCCCCCCCOCc1ccccn1

InChI

1S/C24H43NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-19-22-26-23-24-20-17-18-21-25-24/h17-18,20-21H,2-16,19,22-23H2,1H3

InChI key

CIMKFNVBRHPSRA-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

The membrane for electrode was prepared with 3wt% of this compound 20mol% of K-TCPB , 67.9%Bis(1-butylpentyl)adipate(BBPA) as solvent membrane and 29.1% of PVC.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service