Skip to Content
Merck
All Photos(1)

Documents

18960

Sigma-Aldrich

1,4-Butanediol

purum, ≥98.0% (GC)

Synonym(s):

1,4-Butylene glycol, Tetramethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)4OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1633445
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

3.1 (vs air)

grade

purum

Assay

≥98.0% (GC)

autoignition temp.

698 °F

refractive index

n20/D 1.445 (lit.)
n20/D 1.446

bp

230 °C (lit.)

mp

16 °C (lit.)

density

1.017 g/mL at 25 °C (lit.)

SMILES string

OCCCCO

InChI

1S/C4H10O2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

WERYXYBDKMZEQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

273.2 °F - closed cup

Flash Point(C)

134 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service