Skip to Content
Merck
All Photos(1)

Key Documents

178810-M

Sigma-Aldrich

Tetrahydrofuran

≥99.0%, reagent grade, contains 250 ppm BHT as inhibitor

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Tetrahydrofuran, reagent grade, ≥99.0%, contains 250 ppm BHT as inhibitor

grade

reagent grade

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

610 °F

contains

250 ppm BHT as inhibitor

expl. lim.

1.8-11.8 %

dilution

(for analytical testing)

refractive index

n20/D 1.407 (lit.)

pH

~7

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran (THF) is a saturated cyclic ether mainly used as an organic solvent. On long term storage it forms organic peroxides. This process can be suppressed by adding butylated hydroxytoluene (BHT) as a stabilizer. BHT removes the free radicals required for the peroxide formation. THF constitutes the key fragment of various natural products (polyether antibiotics).[1] THF can form a double hydrate with hydrogen sulfide. Crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies.[2] Butane-1,4-diol is formed as an intermediate during the synthesis of THF.[3] Hot THF is useful for the dissolution of polyvinylidene chloride (PVDV).[4]

Application

Tetrahydrofuran may be used for the dissolution of poly-ε-caprolactone (PCL) and 1,3-diaminopentane, during the preparation of poly-ε-caprolactone (PCL)-hydroxyapatite (HA) scaffolds[5] and acrylate-terminated poly(5-amino-1-pentanol-co-1,4-butanediol diacrylate) (C32)- 1,3-diaminopentane (117) polymer,[6] respectively.

Other Notes

For information on tetrahydrofuran miscibility, please visit the following link:
Tetrahydrofuran Miscibility/Immiscibility Table

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An experimental and modeling study of the combustion of tetrahydrofuran.
Tran LS, et al.
Combustion and Flame, 162(5), 1899-1918 (2015)
Todd J Harris et al.
Biomaterials, 31(5), 998-1006 (2009-10-24)
The use of biomaterials for gene delivery can potentially avoid many of the safety concerns with viral gene delivery. However, the efficacy of polymeric gene delivery methods is low, particularly in vivo. One significant concern is that the interior and
Haiying Yu et al.
Biomaterials, 30(4), 508-517 (2008-11-01)
Natural bone growth greatly depends on the precedent vascular network that supplies oxygen and essential nutrients and removes metabolites. Likewise, it is crucial for tissue-engineered bone to establish a vascular network that temporally precedes new bone formation, and spatially originates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service