Skip to Content
Merck
All Photos(1)

Documents

17310

Sigma-Aldrich

Paraffin wax

purum

Synonym(s):

Paraffins

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

grade

purum

Quality Level

form

crystalline (chunks)
powder or solid

transition temp

solidification point 60-65 °C

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

602.6 °F - Pensky-Martens closed cup

Flash Point(C)

317 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Haohao Zhong et al.
Diagnostic molecular pathology : the American journal of surgical pathology, part B, 22(1), 52-58 (2013-02-02)
We report a hydrothermal pressure method (pressure cooking) for simultaneous deparaffinization and lysis of formalin-fixed paraffin-embedded tissue followed by conventional chaotropic salt column purification to obtain high-quality DNA. Using this method, the release of DNA occurred within the first minute
Wisam Al-Faqheri et al.
PloS one, 8(3), e58523-e58523 (2013-03-19)
This paper introduces novel vacuum/compression valves (VCVs) utilizing paraffin wax. A VCV is implemented by sealing the venting channel/hole with wax plugs (for normally-closed valve), or to be sealed by wax (for normally-open valve), and is activated by localized heating
Abdulazeez Salawu et al.
PloS one, 7(11), e50415-e50415 (2012-12-05)
Most soft tissue sarcomas are characterized by genetic instability and frequent genomic copy number aberrations that are not subtype-specific. Oligonucleotide microarray-based Comparative Genomic Hybridisation (array CGH) is an important technique used to map genome-wide copy number aberrations, but the traditional
Yu Seob Shin et al.
Urology, 81(2), 437-441 (2013-02-05)
To introduce a new repair technique, inverted V-shape, for penile paraffinoma without necrosis of the ventral skin anastomosed. From March 2006 to July 2012, 34 patients underwent penile paraffinoma repair using the T-style or a new operation procedure, the inverted
Roman Zimmermann et al.
Plant physiology, 152(1), 356-365 (2009-11-21)
In an approach to study lateral root development in monocots, genome-wide searches for homologs of the Gibberellic Acid Stimulated Transcript-like (GAST-like) gene family in rice (Oryza sativa) and maize (Zea mays) were carried out. Six novel GAST-like genes in rice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service