Skip to Content
Merck
All Photos(1)

Key Documents

17168

Millipore

Milk Agar, modified according to Brown & Scott

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
bile salts mixture, 1.5 g/L
instant Non-Fat Milk, 100 g/L
peptic digest of animal tissue, 5 g/L
sodium chloride, 5 g/L
yeast extract, 1.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

environmental
food and beverages
water monitoring

microbiology

suitability

selective for Pseudomonas spp.

Application

For the confirmation of Pseudomonas aeruginosa in swimming pool waters.

Components

Part A (Cat. No.22287) 391 g + Part B (Cat. No. 28208) 109g + instruction leaflet

Preparation Note

Part A: Suspend 100 g in 500 ml distilled water. Sterilize by autoclaving at 121°C for 5 minutes. Cool to 55°C.
Part B: Suspend 28 g in 500 ml distilled water and sterilize by autoclaving at 121°C for 15 minutes. Cool rapidly to 55°C.
Mix Part A and Part B together and pour into sterile petri plates.

Legal Information

NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service