Skip to Content
Merck
All Photos(1)

Documents

16171

Sigma-Aldrich

SODOSIL RA 08

alkaline cleaner, with desinfecting effect

Sign Into View Organizational & Contract Pricing


About This Item

UBA Number:
08050037
UNSPSC Code:
12352100

form

solid

composition

active chlorine, 1.0-2.5%
anionic surfactants, <5%
phosphate, 15.0-20.0%

Application

SODOSIL was used as detergent in rinsing epoxy-coated surfaces and multielectrode arrays.

Legal Information

SODOSIL is a trademark of Sigma-Aldrich Chemie GmbH

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Aspenes et al.
Journal of colloid and interface science, 343(2), 529-536 (2009-12-29)
The mechanisms by which hydrates deposit in a petroleum production line are related to pipeline surface properties, fluid composition and properties, and water cut. In this work, adhesion forces between cyclopentane hydrates and solid surfaces were investigated as a function
Shimshon Jacobi et al.
Journal of neurophysiology, 97(5), 3597-3606 (2007-03-09)
We investigate the propagation of neural activity along one-dimensional rat hippocampal cultures patterned in lines over multielectrode arrays. Activity occurs spontaneously or is evoked by local electrical or chemical stimuli, with different resulting propagation velocities and firing rate amplitudes. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service