Skip to Content
Merck
All Photos(1)

Key Documents

15861

Supelco

Citral

analytical standard

Synonym(s):

3,7-Dimethyl-2,6-octadienal, Geranial and neral mixture

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$363.00

R$363.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
R$363.00

About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCHO
CAS Number:
Molecular Weight:
152.23
Beilstein:
1721871
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

R$363.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

5 (vs air)

vapor pressure

0.2 mmHg ( 200 °C)

Assay

≥98.0% (sum of cis and trans, GC)
48.0-52.0% (cis)
48.0-52.0% (trans)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.487-1.489
n20/D 1.488 (lit.)

bp

229 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)C=C(C)CC\C=C(\C)C

InChI

1S/C10H16O/c1-9(2)5-4-6-10(3)7-8-11/h5,7-8H,4,6H2,1-3H3

InChI key

WTEVQBCEXWBHNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Geranial : Neral 1:1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - DIN 51758

Flash Point(C)

98 °C - DIN 51758


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service