Skip to Content
Merck
All Photos(1)

Key Documents

15150

Sigma-Aldrich

1,4-Bis(5-phenyl-2-oxazolyl)benzene

purum, suitable for scintillation, ≥97.0% (TLC)

Synonym(s):

1,4-Bis(5-phenyl-2-oxazolyl)benzene, 2,2′-p-Phenylene-bis(5-phenyloxazole), Di(phenyl-5-oxazolyl-2)-1,4-benzene, POPOP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16N2O2
CAS Number:
Molecular Weight:
364.40
Beilstein:
325741
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

purum

Assay

≥97.0% (TLC)

mp

242-246 °C (lit.)
243-246 °C

suitability

suitable for scintillation

SMILES string

c1ccc(cc1)-c2cnc(o2)-c3ccc(cc3)-c4ncc(o4)-c5ccccc5

InChI

1S/C24H16N2O2/c1-3-7-17(8-4-1)21-15-25-23(27-21)19-11-13-20(14-12-19)24-26-16-22(28-24)18-9-5-2-6-10-18/h1-16H

InChI key

MASVCBBIUQRUKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

POPOP is a fluorescent reagent that has been shown to be phototoxic against several types of microorganisms and, therefore, is capable of producing singlet oxygen . Utilized as a secondary scintillator in scintillation cocktails .

Quantity

λmax. 358 nm, log ε 4.72

replaced by

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Zhao et al.
JOP : Journal of the pancreas, 2(2), 78-82 (2002-02-28)
To develop a method for the primary culture of porcine pancreatic acinar cells. Dispersed pancreatic acinar cells available utilizing RPMI-1640 medium containing collagenase III. After purification, the isolated acinar cells were cultured in RPMI-1640 medium with the addition of 2.5%
Formulation for the preparation of a beta scintillating plastic devicec
Cardarelli, F., Chemla, M., Simonin, J.-P., et al.
Optical Materials, 4, 736-736 (1995)
L A Fothergill et al.
The Biochemical journal, 116(4), 555-561 (1970-02-01)
1. The thiol and disulphide contents of hen ovalbumin were investigated by p-chloromercuribenzoate titration, by determination of cysteic acid content after performic acid oxidation, by measurement of uptake of radioactive iodoacetic acid, and by assay of S-aminoethylcysteine after reaction with
Zhiqi Shi et al.
International journal of molecular sciences, 9(3), 371-382 (2009-03-28)
The effects of osthol, a plant coumarin, on morphology, sugar uptake and cell wall components of Fusarium graminearum were examined in vitro by electron microscopy,(14)C-labelling and enzyme activity detection. The results revealed that osthol could inhibit the hypha growth of
Keisaku Sato et al.
Anesthesiology, 112(6), 1350-1354 (2010-05-13)
Malignant hyperthermia is associated with mutations within the gene encoding the skeletal muscle ryanodine receptor, the calcium channel that releases Ca from sarcoplasmic reticulum stores triggering muscle contraction, and other metabolic activities. More than 200 variants have been identified in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service