Skip to Content
Merck
All Photos(1)

Documents

14389

Supelco

Delphinidin chloride

analytical standard

Synonym(s):

3,3′,4′,5,5′,7-Hexahydroxyflavylium chloride, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-1-benzopyrylium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClO7
CAS Number:
Molecular Weight:
338.70
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤8.0% water

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Cl-].Oc1cc(O)c2cc(O)c([o+]c2c1)-c3cc(O)c(O)c(O)c3

InChI

1S/C15H10O7.ClH/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6;/h1-5H,(H5-,16,17,18,19,20,21);1H

InChI key

FFNDMZIBVDSQFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anthocyanidin. Antioxidant. Pigment in grapes, cranberries, and pomegranates.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Lou et al.
Journal of experimental botany, 65(12), 3157-3164 (2014-05-03)
Grape hyacinth (Muscari) is an important ornamental bulbous plant with an extraordinary blue colour. Muscari armeniacum, whose flowers can be naturally white, provides an opportunity to unravel the complex metabolic networks underlying certain biochemical traits, especially colour. A blue flower
Yue Zhu et al.
Planta, 241(5), 1075-1089 (2015-01-13)
Metabolic profiling, gene cloning, enzymatic analysis, ectopic expression, and gene silencing experiments demonstrate that the anthocyanidin reductase (ANR) pathway is involved in the biosynthesis of proanthocyanidins in upland cotton. Proanthocyanidins (PAs) are oligomeric or polymeric flavan-3-ols, however, the biosynthetic pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service