Skip to Content
Merck
All Photos(1)

Key Documents

12926

Sigma-Aldrich

Cobalt(III) oxide

purum, ≥70% Co basis, black

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co2O3
CAS Number:
Molecular Weight:
165.86
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

grade

purum

Assay

≥70% Co basis

form

powder

color

black

cation traces

Fe: ≤1000 mg/kg
Ni: ≤3000 mg/kg
Pb: ≤200 mg/kg

SMILES string

O=[Co]O[Co]=O

InChI

1S/2Co.3O

InChI key

UPWOEMHINGJHOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics and reaction stoichiometry in the reductive dissolution of manganese (IV) dioxide and Co (III) oxide by hydroquinone.
Stone AT and Ulrich H-J.
Journal of Colloid and Interface Science, 132(2), 509-522 (1989)
Wet oxidation of oxygen-and nitrogen-containing organic compounds catalyzed by cobalt (III) oxide.
Ito MM, et al.
Industrial & Engineering Chemistry Research, 28(7), 894-899 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service