Skip to Content
Merck
All Photos(1)

Documents

12288

Supelco

Metalaxyl-(phenyl-13C6)

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C9H21NO4
CAS Number:
Molecular Weight:
285.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

69-73 °C

format

neat

mass shift

M+6

storage temp.

−20°C
−20°C

SMILES string

COCC(=O)N(C(C)C(=O)OC)[13c]1[13c](C)[13cH][13cH][13cH][13c]1C

InChI

1S/C15H21NO4/c1-10-7-6-8-11(2)14(10)16(13(17)9-19-4)12(3)15(18)20-5/h6-8,12H,9H2,1-5H3/i6+1,7+1,8+1,10+1,11+1,14+1

InChI key

ZQEIXNIJLIKNTD-HEYPXIGASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metalaxyl-(phenyl-13C6) is an isotope-labeled analog of the phenylamide fungicide, metalaxyl.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective degradation kinetics of metalaxyl in rabbits
Qiu J, et al.
Pesticide Biochemistry and Physiology, 83(1), 1-8 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service