Skip to Content
Merck
All Photos(1)

Key Documents

12228

Sigma-Aldrich

Chromium(III) chloride hexahydrate

purum, ≥96%

Synonym(s):

Chromium trichloride hexahydrate, Hexaaquachromium(III) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrCl3 · 6H2O
CAS Number:
Molecular Weight:
266.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥96%

form

solid

reaction suitability

core: chromium
reagent type: catalyst

density

1.76 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤1000 mg/kg

cation traces

Fe: ≤300 mg/kg

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Cr](Cl)Cl

InChI

1S/3ClH.Cr.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

LJAOOBNHPFKCDR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongqing Li et al.
Journal of immunology (Baltimore, Md. : 1950), 202(4), 1219-1228 (2019-01-16)
The appropriate inflammatory response is essential for normal wound repair, and skin commensal Staphylococcus epidermidis has been shown to regulate TLR3-mediated inflammatory response to maintain skin homeostasis after injury. However, the underlying mechanism by which S. epidermidis regulates wound-induced inflammation
Arun Balasubramaniam et al.
Scientific reports, 10(1), 21585-21585 (2020-12-11)
Repurposing existing compounds for new indications may facilitate the discovery of skin prebiotics which have not been well defined. Four compounds that have been registered by the International Nomenclature of Cosmetic Ingredients (INCI) were included to study their abilities to
Sunita Keshari et al.
International journal of molecular sciences, 20(18) (2019-09-14)
The glycerol fermentation of probiotic Staphylococcus epidermidis (S. epidermidis) in the skin microbiome produced butyric acid in vitro at concentrations in the millimolar range. The exposure of dorsal skin of mice to ultraviolet B (UVB) light provoked a significant increased
Shinta Marito et al.
International journal of molecular sciences, 21(14) (2020-07-28)
The probiotic activity of skin Staphylococcus epidermidis (S. epidermidis) bacteria can elicit diverse biological functions via the fermentation of various carbon sources. Here, we found that polyethylene glycol (PEG)-8 Laurate, a carbon-rich molecule, can selectively induce the fermentation of S.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service