11230
L-Aspartic acid potassium salt
puriss. p.a., ≥99.0% (anhydrous basis)
Synonym(s):
Potassium L-aspartate
About This Item
Recommended Products
grade
puriss. p.a.
Quality Level
Assay
≥99.0% (T)
≥99.0% (anhydrous basis)
form
powder
optical activity
[α]20/D −13.5±1°, c = 1% in H2O
impurities
≤6% water
color
white
anion traces
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤500 mg/kg
cation traces
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
Mg: ≤100 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤250 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
SMILES string
N[C@H](C([O-])=O)CC(O)=O.[K+]
InChI
1S/C4H7NO4.K/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H,6,7)(H,8,9);/q;+1/p-1/t2-;/m0./s1
InChI key
TXXVQZSTAVIHFD-DKWTVANSSA-M
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service