Skip to Content
Merck
All Photos(1)

Documents

109665

Sigma-Aldrich

1,3-Diphenyltriazene

95%

Synonym(s):

Diazoaminobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHN=NC6H5
CAS Number:
Molecular Weight:
197.24
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

vapor density

6.8 (vs air)

Assay

95%

bp

146 °C (lit.)

SMILES string

N(\N=N\c1ccccc1)c2ccccc2

Application

Diazoaminobenzene is an azo compound used as an intermediate, complexing agent, and polymer additive. DAAB is a manufacturing intermediate that metabolizes primarily to known carcinogens, benzene and aniline. DAAB has been identified as an impurity in a number of dyes and coloring agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diazoaminobenzene.
Report on carcinogens : carcinogen profiles, 12, 132-134 (2011-08-19)
Feng Li et al.
Biosensors & bioelectronics, 25(9), 2084-2088 (2010-03-09)
A novel protocol for the gold nanoparticles (AuNPs) modification on the electrode surface was proposed, which was based on the self-assembly of AuNPs on the mercapto-diazoaminobenzene monolayer modified electrode. The mercapto-diazoaminobenzene monolayer was obtained by covalent immobilization of 4-aminothiophenol (4-ATP)
Chhanda Mukhopadhyay et al.
Organic & biomolecular chemistry, 8(20), 4720-4729 (2010-09-08)
An efficient and highly versatile method for the synthesis of diverse regiospecific 1-arylbenzotriazoles being important medicinal scaffolds, by the copper (1) catalysed intramolecular N-arylation of diazoaminobenzenes of 2-haloaryldiazonium salts in PEG-water has been developed. A very simple reaction protocol, large
J E Bailey
Journal of chromatography, 321(1), 185-197 (1985-03-08)
Data are presented for the determination of trace levels of 1,3-diphenyltriazene (DPT) and azobenzene (AB) in D&C Red No. 33. The contaminants are extracted with chloroform from an aqueous solution of the color, and the chloroform is removed under vacuum.
Nancy B Ress et al.
Mutation research, 521(1-2), 201-208 (2002-11-20)
Diazoaminobenzene (DAAB), a manufacturing intermediate metabolized primarily to the known carcinogens benzene and aniline, has been identified as an impurity in a number of dyes and coloring agents that are components of cosmetics, food products, and pharmaceuticals. Several structural analogs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service