Skip to Content
Merck
All Photos(1)

Key Documents

10952

Sigma-Aldrich

Araldite® M accelerator 960

accelerator DY 060

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

Quality Level

refractive index

n/D 1.517

application(s)

hematology
histology

storage temp.

room temp

General description

Araldite is an epoxy resin commonly used for embedding. It is basically free from polymerization damage and the plastic does not sublime in the microscope, hence providing continual support and preservation of fine detail.[1] It is composed of araldite M or CY 212, dodecenyl succinic anhydride, dibutyl phthalate and 2,4,6-tridimethylamine methyl phenol.[2]

Application

Araldite® was used to study electrochemical detection of dopamine and ascorbic acid neurotransmitters focusing on miniaturization of a graphite-epoxy and synthetic zeolite-modified graphite-epoxy composite macroelectrode as a quasi-microelectrode.[3] It was also used in production of microfluidic chips using extremely low cost chemicals and equipment which was validated by producing monodisperse polymeric microparticles for drug delivery and hydrogel microfibers for cell embedding.[4]

Legal Information

Araldite is a registered trademark of Huntsman Advanced Materials Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid M Parsons et al.
Dalton transactions (Cambridge, England : 2003), 48(29), 10945-10952 (2019-06-06)
The active fragment [Tp'Rh(CNneopentyl)], generated from the precursor Tp'Rh(CNneopentyl)(PhN[double bond, length as m-dash]CNneopentyl), underwent oxidative addition of substituted ketones and esters resulting in Tp'Rh(CNneopentyl)(R)(H) complexes (Tp' = tris-(3,5-dimethylpyrazolyl)borate). These C-H activated complexes underwent reductive elimination at varying temperatures (24-70 °C)
S Focaroli et al.
Lab on a chip, 14(20), 4007-4016 (2014-08-22)
The present paper describes the production of microfluidic chips using an approach based on shrinkable biocompatible polymers (i.e. agarose) for the production of size controlled microfluidic channels. In addition, all steps of chip production were carried out using an inexpensive
Simultaneous/selective detection of dopamine and ascorbic acid at synthetic zeolite-modified/graphite-epoxy composite macro/quasi-microelectrodes.
Ilinoiu EC
Sensors, 13 (6), 7296-7307 (2013)
In Situ Hybridization in Electron Microscopy.
Morel G
Science, 174-175 (2001)
Biological Techniques.
Yadav P.R.
Science, 79-80 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service