Skip to Content
Merck
All Photos(1)

Key Documents

10832

Sigma-Aldrich

2-Phenoxyethanol

tested according to Ph. Eur.

Synonym(s):

Phenoxyethanolum, Ethylene glycol monophenyl ether, Phenylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
1364011
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

form

liquid

bp

244-246 °C

mp

11-13 °C

density

1.107 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

OCCOc1ccccc1

InChI

1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

QCDWFXQBSFUVSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao-Ping Weng et al.
Reproductive toxicology (Elmsford, N.Y.), 29(4), 452-457 (2010-04-13)
Ethylene glycol monomethyl ether (EGME) is known to alter the reproductive function of exposed animals and their offspring; however, its influence on cumulus-oocyte complexes (COCs) remains unclear. This study assessed the toxicity of EGME on oocytes and COCs by analyzing
Santosh K Tripathi et al.
Organic & biomolecular chemistry, 9(2), 581-587 (2010-11-05)
Syntheses of several diorganodiselenides and, in particular, a seven-membered cyclic seleninate ester derived from 2-phenoxyethanol are described. The seleninate ester was obtained from allyl (2-(2-hydroxyethoxy)phenyl) selenide through a series of oxidation and [2,3] sigmatropic rearrangement steps. The ester exhibits good
Jessica Stahl et al.
International wound journal, 7(1), 62-69 (2010-04-23)
A combination of 0.1% octenidine dihydrochloride and 2% 2-phenoxyethanol (octenisept) is a commonly used disinfectant in human medicine. As porcine skin represents an adequate model for human skin, the effect of octenidine dihydrochloride and phenoxyethanol on wound healing is studied
J Lademann et al.
Skin pharmacology and physiology, 25(2), 100-106 (2012-02-04)
Colonization and infection of wounds represent a major reason for the impairment of tissue repair. Recently, it has been reported that tissue-tolerable plasma (TTP) is highly efficient in the reduction of the bacterial load of the skin. In the present
Tirumurugaan Krishnaswamy Gopalan et al.
PloS one, 9(6), e100018-e100018 (2014-06-24)
From an immunologist perspective, sharks are an important group of jawed cartilaginous fishes and survey of the public database revealed a great gap in availability of large-scale sequence data for the group of Chondrichthyans the elasmobranchs. In an attempt to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service