Skip to Content
Merck
All Photos(1)

Key Documents

06416

Supelco

Aluminum silicate

washed, fibers (may contain particles)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
47131900
PubChem Substance ID:
NACRES:
NA.21

form

fibers (may contain particles)

quality

washed

technique(s)

LPLC: suitable

impurities

asbestos, none detected

matrix

Aluminum silicate

matrix active group

aluminum

separation technique

normal phase

InChI

1S/6Al.2O2Si.9O/c;;;;;;2*1-3-2;;;;;;;;;

InChI key

KZHJGOXRZJKJNY-UHFFFAOYSA-N

Analysis Note

classification-temperature: 1260°C (the classification-temperature has been detected by irreversible, linear change, not by melting point)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service