Skip to Content
Merck
All Photos(1)

Key Documents

05295001

Hypericum perforatum extract

secondary reference standard

Synonym(s):

St. John´s wort extract

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$1,925.00

R$1,925.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$1,925.00

About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,925.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

matrix material

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Hypericum perforatum L., herba extract (according to HMPC Monograph)

genus hypericum
Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: Hypericin (quantitative); Pseudohypericin (qualitative)

Matrix Group: Botanicals

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service