Skip to Content
Merck
All Photos(1)

Key Documents

04930

Sigma-Aldrich

N-Ethyl-m-toluidine

≥98.0% (GC)

Synonym(s):

3-(Ethylamino)toluene, N-Ethyl-3-aminotoluene, N-Ethyl-3-methylaniline

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
R$843.00

R$843.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
R$843.00

About This Item

Linear Formula:
CH3C6H4NHC2H5
CAS Number:
Molecular Weight:
135.21
Beilstein:
742170
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
technique(s):
GC/GC: suitable
bp:
221 °C (lit.)

R$843.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

technique(s)

GC/GC: suitable

refractive index

n20/D 1.546 (lit.)
n20/D 1.546

bp

221 °C (lit.)

density

0.957 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCNc1cccc(C)c1

InChI

1S/C9H13N/c1-3-10-9-6-4-5-8(2)7-9/h4-7,10H,3H2,1-2H3

InChI key

GUYMMHOQXYZMJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Direct 18F-Fluorosulfurylation of Phenols and Amines Using an FSO2+ Transfer Agent Generated In Situ: Reports the synthesis process involving N-ethyl-m-toluidine as a reagent in the preparation of the title compound for fluorosulfurylation applications (MP Kim, H Cho, S Kayal, MH Jeon, JK Seo, 2023).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service