Skip to Content
Merck
All Photos(1)

Key Documents

04542

Sigma-Aldrich

(S)-Boc-3,4-dimethoxy-β-Phe-OH

≥98.0% (HPLC)

Synonym(s):

(S)-3-(Boc-amino)-3-(3,4-methoxyphenyl)propionic acid, Boc-3,4-dimethoxy-D-β-phenylalanine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$3,084.00

R$3,084.00


Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
R$3,084.00

About This Item

Empirical Formula (Hill Notation):
C16H23NO6
CAS Number:
Molecular Weight:
325.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

R$3,084.00


Estimated to ship onMay 31, 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

powder

color

white

mp

142-146 °C

application(s)

peptide synthesis

SMILES string

COc1ccc(cc1OC)[C@H](CC(O)=O)NC(=O)OC(C)(C)C

InChI

1S/C16H23NO6/c1-16(2,3)23-15(20)17-11(9-14(18)19)10-6-7-12(21-4)13(8-10)22-5/h6-8,11H,9H2,1-5H3,(H,17,20)(H,18,19)/t11-/m0/s1

InChI key

NOEVPXYRCVCOIV-NSHDSACASA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service