04420
Phosphinic acid
puriss., meets analytical specification of NF, 30-32%
Synonym(s):
Hypophosphorous acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
puriss.
quality
meets analytical specification of NF
concentration
30-32%
impurities
≤0.002% heavy metals (as Pb)
density
1.206 g/mL at 20 °C (lit.)
anion traces
chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤200 mg/kg
cation traces
As: ≤1 mg/kg
Ca: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
Na: ≤1000 mg/kg
SMILES string
O[PH2]=O
InChI
1S/H3O2P/c1-3-2/h3H2,(H,1,2)
InChI key
ACVYVLVWPXVTIT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Phosphinic acid is a reagent containing P-H bond. Phosphinic acids (phosphinates) are the derivatives of phosphinic acid H2P(O)(OH). Biochemical, biological, and medical applications of phosphinic acid compounds have been evaluated. Natural products containing carbon-phosphorus bonds (phosphonic and phosphinic acids) have wide applications in medicine and agriculture. Phosphinic acids have been synthesized via Michael addition of phosphonous acids as well as esters to activated conjugated systems. Phosphinic acid (Hypophosphorous acid) is an effective radical reducing agent for organic halides, thionoesters and isocyanides.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Preparation of phosphinic acids: Michael additions of phosphonous acids/esters to conjugated systems.
Tetrahedron Letters, 25(42), 4741-4744 (1984)
Annual review of biochemistry, 78, 65-94 (2009-06-06)
Natural products containing carbon-phosphorus bonds (phosphonic and phosphinic acids) have found widespread use in medicine and agriculture. Recent years have seen a renewed interest in the biochemistry and biology of these compounds with the cloning of the biosynthetic gene clusters
Hypophosphorous acid and its salts: New reagents for radical chain deoxygenation, dehalogenation and deamination.
Tetrahedron Letters, 33(39), 5709-5712 (1992)
Current medicinal chemistry, 7(6), 629-647 (2000-03-07)
This review summarizes our knowledge of biochemical, biological and medical applications and properties of phosphinic acid compounds. Phosphinic acid compounds (phosphinates) are derivatives of phosphinic acid H2P(O)(OH). The major attention of this article is focused on applications of phosphinates of
The invention of radical reactions. Part 32. Radical deoxygenations, dehalogenations, and deaminations with dialkyl phosphites and hypophosphorous acid as hydrogen sources.
The Journal of Organic Chemistry, 58(24), 6838-6842 (1993)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service