04150
Iodoethane
puriss., ≥99.5% (GC)
Synonym(s):
Ethyl iodide
About This Item
Recommended Products
vapor density
5.38 (vs air)
vapor pressure
100 mmHg ( 18 °C)
grade
puriss.
Assay
≥99.5% (GC)
contains
silver wool as stabilizer
refractive index
n20/D 1.513 (lit.)
n20/D 1.514
bp
69-73 °C (lit.)
mp
−108 °C (lit.)
density
1.940 g/mL at 25 °C (lit.)
SMILES string
CCI
InChI
1S/C2H5I/c1-2-3/h2H2,1H3
InChI key
HVTICUPFWKNHNG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
127.4 °F - closed cup - (External MSDS)
Flash Point(C)
53 °C - closed cup - (External MSDS)
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service