Skip to Content
Merck
All Photos(1)

Key Documents

03830

Sigma-Aldrich

Ethylene glycol monosalicylate

≥98.0% (GC)

Synonym(s):

Ethylene glycol salicylate, Salicylic acid 2-hydroxyethyl ester, Salicylic acid ethylene glycol ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
R$584.00

R$584.00


Available to ship onApril 01, 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
R$584.00

About This Item

Linear Formula:
HOC6H4COOCH2CH2OH
CAS Number:
Molecular Weight:
182.17
Beilstein:
2255685
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$584.00


Available to ship onApril 01, 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.548 (lit.)
n20/D 1.549

bp

166 °C/13 mmHg (lit.)

density

1.244 g/mL at 25 °C (lit.)

SMILES string

OCCOC(=O)c1ccccc1O

InChI

1S/C9H10O4/c10-5-6-13-9(12)7-3-1-2-4-8(7)11/h1-4,10-11H,5-6H2

InChI key

LVYLCBNXHHHPSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

339.8 °F - DIN 51758

Flash Point(C)

171 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact dermatitis from a veterinary antiinflammatory gel containing 2-hydroxyethyl salicylate.
Susana Córdoba et al.
Dermatitis : contact, atopic, occupational, drug, 22(3), 171-172 (2011-05-17)
Contact dermatitis from hydroxyethyl salicylate.
C Reichert et al.
Contact dermatitis, 33(4), 275-276 (1995-10-01)
Miroslav Kucera et al.
Arzneimittel-Forschung, 53(12), 850-856 (2004-01-22)
A combination of the active agents arnica and hydroxyethyl salicylate (HES) in ethanolic solution (Sportino Acute Spray) is cutaneously applied for the treatment of sports injuries and diseases of the locomotor apparatus. The aim was to examine the efficacy and
Contact dermatitis from anti-inflammatory gel containing hydroxyethyl salicylate.
J Horak et al.
Contact dermatitis, 47(2), 120-121 (2002-11-29)
S E Cross et al.
British journal of clinical pharmacology, 46(1), 29-35 (1998-08-05)
The penetration of active ingredients from topically applied anti-inflammatory pharmaceutical products into tissues below the skin is the basis of their therapeutic efficacy. There is still controversy as to whether these agents are capable of direct penetration by diffusion through

Questions

  1. Hi Can we use ( product code- 03830) reagent grade as a reference standard?

    1 answer
    1. This product is intended for general laboratory research use and has not been qualified for use as an analytical standard. Please see the links below to review products that are suitable for use as standards:
      PHL80106
      https://www.sigmaaldrich.com/product/supelco/phl80106
      H1429400
      https://www.sigmaaldrich.com/product/sial/h1429400

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service