Skip to Content
Merck
All Photos(1)

Key Documents

02672

Supelco

Calcium atomic spectroscopy standard concentrate 1.00 g Ca

1.00 g/L, for 1L standard solution, analytical standard

Synonym(s):

Calcium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2
CAS Number:
Molecular Weight:
110.98
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

packaging

ampule of

concentration

1.00 g/L

solubility

H2O: soluble

SMILES string

Cl[Ca]Cl

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Preparation Note

prepared with CaCl2, HCl and H2O

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Levi et al.
Journal of thrombosis and haemostasis : JTH, 12(9), 1428-1436 (2014-05-09)
Four-factor prothrombin complex concentrates (PCCs), which contain factor II, FVII, FIX, and FX, have shown the potential to reverse the anticoagulant effect of rivaroxaban in healthy volunteers. The purpose of this study was to determine whether a three-factor PCC, which contains
J Philippe et al.
International journal of obesity (2005), 39(2), 295-302 (2014-06-04)
A significant proportion of severe familial forms of obesity remain genetically elusive. Taking advantage of our unique cohort of multigenerational obese families, we aimed to assess the contribution of rare mutations in 29 common obesity-associated genes to familial obesity, and
Meral Tunc-Ozdemir et al.
Plant physiology, 161(2), 1010-1020 (2013-02-02)
Cyclic nucleotide-gated channels (CNGCs) have been implicated in diverse aspects of plant growth and development, including responses to biotic and abiotic stress, as well as pollen tube growth and fertility. Here, genetic evidence identifies CNGC16 in Arabidopsis (Arabidopsis thaliana) as
Michael A Goren et al.
Nature communications, 5, 5115-5115 (2014-10-09)
Opsin, the rhodopsin apoprotein, was recently shown to be an ATP-independent flippase (or scramblase) that equilibrates phospholipids across photoreceptor disc membranes in mammalian retina, a process required for disc homoeostasis. Here we show that scrambling is a constitutive activity of
Mohsen Akbari et al.
Advanced functional materials, 24(26), 4060-4067 (2014-11-21)
The fabrication of cell-laden structures with anisotropic mechanical properties while having a precise control over the distribution of different cell types within the constructs is important for many tissue engineering applications. Automated textile technologies for making fabrics allow simultaneous control

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service