Skip to Content
Merck
All Photos(1)

Key Documents

02643

Sigma-Aldrich

8-Ethoxy-5-quinolinesulfonic acid sodium salt monohydrate

≥97.0%

Synonym(s):

Sodium 8-ethoxyquinoline-5-sulfonate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10NNaO4S · H2O
CAS Number:
Molecular Weight:
293.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0%

SMILES string

[Na+].CCOc1ccc(c2cccnc12)S([O-])(=O)=O

InChI

1S/C11H11NO4S.Na.H2O/c1-2-16-9-5-6-10(17(13,14)15)8-4-3-7-12-11(8)9;;/h3-7H,2H2,1H3,(H,13,14,15);;1H2/q;+1;/p-1

InChI key

BSVDXMZRSQZXAH-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shampur Narayan Madhusudana et al.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases, 8(6), 339-345 (2004-10-21)
The presently advocated tests for rapid diagnosis of rabies, such as the fluorescent antibody test (FAT) are expensive and require expertise to carry out and interpret the results. In this study, a simple direct dot blot enzyme immunoassay (DIA) has
A Yayon et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(22), 10643-10647 (1993-11-15)
Basic fibroblast growth factor (bFGF) is known to bind to its cell-surface receptors with high affinity and in a heparin-dependent manner. In an attempt to predict the receptor recognition site on bFGF we screened phage-epitope libraries with monoclonal antibodies DG2
M Balass et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(22), 10638-10642 (1993-11-15)
Monoclonal antibody (mAb) 5.5 is directed against the ligand-binding site of the nicotinic acetylcholine receptor. The epitope for this antibody is conformation-dependent, and the antibody does not react with synthetic peptides derived from the receptor sequence. We have identified a
Veroniqa Lundbäck et al.
Diabetologia, 61(5), 1112-1123 (2018-03-01)
By genome-wide association meta-analysis, 17 genetic loci associated with fasting serum insulin (FSI), a marker of systemic insulin resistance, have been identified. To define potential culprit genes in these loci, in a cross-sectional study we analysed white adipose tissue (WAT)
Leonhard Möckl et al.
Developmental cell, 50(1), 57-72 (2019-05-21)
The mammalian glycocalyx is a heavily glycosylated extramembrane compartment found on nearly every cell. Despite its relevance in both health and disease, studies of the glycocalyx remain hampered by a paucity of methods to spatially classify its components. We combine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service