Skip to Content
Merck
All Photos(1)

Key Documents

02580

Sigma-Aldrich

O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N′,N′-tetramethyluronium tetrafluoroborate

≥98.0% (CHN)

Synonym(s):

N-[[[(1-Cyano-2-ethoxy-2-oxoethylidene)amino]oxy](dimethylamino)methylene]-N-methyl-methanaminium tetrafluoroborate, TOTU

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O2CC(CN)=NOC[N(CH3)2]=N(CH3)2BF4
CAS Number:
Molecular Weight:
328.07
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (CHN)

reaction suitability

reaction type: Coupling Reactions

mp

144-145 °C
145 °C (dec.) (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.CCOC(=O)\C(=N/O\C(N(C)C)=[N+](/C)C)C#N

InChI

1S/C10H17N4O3.BF4/c1-6-16-9(15)8(7-11)12-17-10(13(2)3)14(4)5;2-1(3,4)5/h6H2,1-5H3;/q+1;-1/b12-8-;

InChI key

FPQVGDGSRVMNMR-JCTPKUEWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
Watson-Crick pairing of oligonucleotides
Esterification of nucleosides to solid phase supports
Reagent for:
Synthesis of selective integrin avβ3 antagonists
Design and synthesis of double stranded DNA libraries
Preparation of conformationally contrained β-amino acid derivatives
Synthesis and application of methyleneoxy pseudopeptide building blocks

Other Notes

New uronium-based coupling reagent for peptide synthesis suitable for critical cases

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Konig et al. et al.
Proc. 21st Eur. Pept. Symp.: Peptides 1990 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service