Skip to Content
Merck
All Photos(1)

Key Documents

02554

Supelco

Fluorescence indicator green 254 nm

suitable for thin layer chromatography (TLC)

Synonym(s):

Zinc silicate, manganese-doped

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

form

powder

technique(s)

thin layer chromatography (TLC): suitable

fluorescence

(shows a green fluorescence when illuminated with UV light (254 nm))

SMILES string

[Zn++].[Zn++].[O-][Si]([O-])([O-])[O-]

InChI

1S/O4Si.2Zn/c1-5(2,3)4;;/q-4;2*+2

InChI key

ZOIVSVWBENBHNT-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service