Skip to Content
Merck
All Photos(1)

Documents

01318

Millipore

Enterococcus faecium Selective Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Enterococcus spp.
Enterococcus spp.

General description

Enterococcus faecium Selective Supplement is used for selective isolation and detection of Enterococcus faecium. Aztreonam present in the supplement inhibits gram-negative bacteria, including Pseudomonas aeruginosa. The cephalexin inhibits most gram-positive bacteria but not Enterococcus species. The supplement is also recommended to differentiate Enterococcus faecium from Enterococcus faecalis

Application

Enterococcus faecium Selective Supplement is used for selective isolation and detection of Enterococcus faecium in food and beverage samples.

Components

(per vial, sufficient for 500 ml medium)
Aztreonam 37.5mg
Cephalexin 25mg

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service