Skip to Content
Merck
All Photos(1)

Key Documents

0120635

Millipore

Maximum Recovery Diluent in bags

suitable for microbiology

Synonym(s):

MRD

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 3 L
R$318.00

R$318.00


Check Cart for Availability

Request more information

Select a Size

Change View
4 X 3 L
R$318.00

About This Item

UNSPSC Code:
41171610
NACRES:
NA.74

R$318.00


Check Cart for Availability

Request more information

shelf life

limited shelf life, expiry date on the label

Quality Level

composition

sodium chloride, 8.5 g/L
tryptone, 1.0 g/L

pH

7.0±0.2

application(s)

microbiology

General description

This medium can be adjusted and/or supplemented according to the performance criteria required.

Application

Maximum Recovery Diluent (MDR) is a diluent for the preparation of stock suspensions and dilutions. It is a protective and isotonic medium used for maximal recovery of microorganisms from a variety of sources. The low concentration of peptone does not cause multiplication of the organisms within 1-2 hours of dilution of the sample.
MDR, also called Tryptone salt broth, is compliance with ISO standards 6887-1 to 5 for the preparation and dilutions of stock suspensions from most of food products.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service