Skip to Content
Merck
All Photos(1)

Key Documents

00563

Millipore

MM ChromoSelect Agar

NutriSelect® Plus, suitable for microbiology

Synonym(s):

ChromoSelect Miller and Mallinson Agar, MM Agar ChromoSelect

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
R$871.00

R$871.00


Estimated to ship onJune 02, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
500 G
R$871.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

R$871.00


Estimated to ship onJune 02, 2025


Request a Bulk OrderRequest more information

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
D-cellobiose, 3.0 g/L
chromogenic mixture, 6.6 g/L
lactose, 10.0 g/L
D-mannitol, 1.2 g/L
peptone (vegetable), 10.0 g/L
D-trehalose, 1.33 g/L
vegetable extract, 2.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.6±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
pathogen testing

microbiology

storage temp.

15-25°C

suitability

nonselective and differential for Citrobacter spp.
nonselective and differential for Escherichia coli
nonselective and differential for Salmonella spp.
nonselective and differential for coliforms
nonselective and differential for enterobacteriaceae

Application

MM ChromoSelect Agar is recommended for identification and differentiation of Salmonella and non-salmonella like Citrobacter from water samples.

Preparation Note

Suspend 49.13 g in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. DO NOT AUTOCLAVE OR OVERHEAT. Cool to 45-50°C and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service