Skip to Content
Merck
All Photos(1)

Key Documents

00233

Sigma-Aldrich

Fmoc-Sar-OPfp

≥96.0%

Synonym(s):

Fmoc-sarcosine pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16F5NO4
CAS Number:
Molecular Weight:
477.38
Beilstein:
7452048
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥96.0%

form

crystalline

color

white

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CN(CC(=O)Oc1c(F)c(F)c(F)c(F)c1F)C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C24H16F5NO4/c1-30(10-17(31)34-23-21(28)19(26)18(25)20(27)22(23)29)24(32)33-11-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,16H,10-11H2,1H3

InChI key

PJYXJFOLTCMMLK-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service