Skip to Content
Merck
All Photos(1)

Documents

00083

Sigma-Aldrich

Boc-Tyr(PO3Me2)-OH

≥96.0% (HPLC)

Synonym(s):

Nα-Boc-O-(dimethylphospho)-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24NO8P
CAS Number:
Molecular Weight:
389.34
Beilstein:
3632773
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.0% (HPLC)

form

solid

impurities

≤25 % (w/w) dioxane

color

white

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

COP(=O)(OC)Oc1ccc(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)cc1

InChI

1S/C16H24NO8P/c1-16(2,3)24-15(20)17-13(14(18)19)10-11-6-8-12(9-7-11)25-26(21,22-4)23-5/h6-9,13H,10H2,1-5H3,(H,17,20)(H,18,19)/t13-/m0/s1

InChI key

SPVKDJVZEGSJSS-ZDUSSCGKSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service