Skip to Content
Merck
All Photos(1)

Documents

30-3460

Sigma-Aldrich

p-Toluidine

SAJ special grade, ≥99.0%

Synonym(s):

4-Aminotoluene, 4-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
471281
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

SAJ special grade

vapor density

3.9 (vs air)

Assay

≥99.0%

form

crystalline

autoignition temp.

899 °F

expl. lim.

6.6 %

availability

available only in Japan

pH

7.8 (7 g/L)

bp

200 °C (lit.)

mp

41-46 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(N)cc1

InChI

1S/C7H9N/c1-6-2-4-7(8)5-3-6/h2-5H,8H2,1H3

InChI key

RZXMPPFPUUCRFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Sens. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutagenic potential of some chemical components of dental materials.
E G Miller et al.
Dental materials : official publication of the Academy of Dental Materials, 2(4), 163-165 (1986-08-01)
Paul G Stevenson et al.
Journal of chromatography. A, 1218(45), 8255-8263 (2011-10-11)
Several simple techniques are presented for the identification of the boundaries of chromatographic peaks. These methods provide a significant reduction in the time needed to perform the rapid, automatic calculation of the central peak moments and to evaluate the quality
Shunji Ito et al.
The Journal of organic chemistry, 70(6), 2285-2293 (2005-03-12)
[reaction: see text] N,N-Di(6-azulenyl)-p-toluidine (1a) and N,N,N',N'-tetra(6-azulenyl)-p-phenylenediamine (2a) and their derivatives with 1,3-bis(ethoxycarbonyl) substituents on each 6-azulenyl group (1b and 2b) were prepared by Pd-catalyzed amine azulenylation and characterized as a study into new aromatic amines for multistage amphoteric redox
D R Doerge et al.
Chemical research in toxicology, 4(5), 556-560 (1991-09-01)
The metabolism of three arylamine substrates by H2O2 in the presence of each of the peroxidative enzymes chloroperoxidase (CPX) and pea seed peroxygenase (PSM) was conducted with normal H2O2 and with 18O-labeled H2O2. The resulting C-nitroso aromatic metabolites were examined
W J Brock et al.
Toxicology letters, 54(2-3), 317-325 (1990-12-01)
The in vivo covalent binding of ortho- and para-toluidine (OT and PT) to rat hepatic macromolecules was investigated to determine if a relationship exists between the degree of binding for each isomer and its carcinogenic potency. The ortho-isomer has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service