Skip to Content
Merck
All Photos(1)

Key Documents

30-3240

Sigma-Aldrich

o-Tolidine solution

Synonym(s):

Ellms-Hauser solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2
CAS Number:
Molecular Weight:
212.29
Beilstein:
2210640
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

availability

available only in Japan

concentration

~0.1 % (w/v)

dilution

(for analytical testing)

SMILES string

Cc1cc(ccc1N)-c2ccc(N)c(C)c2

InChI

1S/C14H16N2/c1-9-7-11(3-5-13(9)15)12-4-6-14(16)10(2)8-12/h3-8H,15-16H2,1-2H3

InChI key

NUIURNJTPRWVAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the determination of chlorine, chloride, gold, etc[1][2][3][4]

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Met. Corr. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.A. Momin et al.
Analytica Chimica Acta, 244, 71-71 (1991)
Stabilized neutral orthotolidine, SNORT, colorimetric method for chlorine.
J D Johnson et al.
Analytical chemistry, 41(13), 1744-1750 (1969-11-01)
B. Demirata et al.
Analytical Sciences, 8, 225-225 (1992)
E. Scheubeck et al.
Z. Anal. Chem., 249, 370-370 (1970)
Shinpei Kawarai et al.
The Journal of veterinary medical science, 72(2), 131-140 (2009-11-27)
It is essential to develop a technique to culture purified skin-derived mast cells (SMCs) to facilitate immunological research on allergic diseases in dogs. This study was performed to develop an efficient culture system for canine SMCs and to characterize the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service